Learn More
Brady™ Paper Labels for M6/M7 Printers
Paper Labels with Permanent Rubber Adhesive provide good contrast and smear resistance for thermal transfer and barcode printing applications.
Brand:  Brady™ 173872
 View more versions of this product
Product Code. 17821629
  • ONLINE EXCLUSIVE PRICE
    £40.05
    £38.20 / Pack of 100
    Save £1.85
    4% Off
In Stock
Add to basket
Description
Description
  • White matte paper label stock features permanent rubber adhesive
  • Low cost is ideal for all general purpose labeling projects
  • Service temperature of -40°C to 50°C (-40°F to 122°F)
  • Made with B-424 permanent adhesive
Specifications
Specifications
Not Autoclavable
Permanent Rubber
White
Die-cut
Single-side Printable
Label
Barcode Labeling, Electrical Labeling, General Identification, Laboratory Labeling
No
Rectangle
-40°C to +50°C
0.089 mm
1.9 in.
101.6 mm
M6-38-424
ASTM D 1000
None
Matte
Paper
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
100 Labels
-40°F to +122°F
0.0035 in.
1.9 x 4 in.
4 in.
48.26 mm