missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAPD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560177
This item is not returnable.
View return policy
Description
Immunogen sequence: QSSSSDVDSD ATPCKTPKQL LCRPSTGNRV GSQDVSLESS QAVGKMQSTQ TTNTSNSTNK SQHGSARLFR SSSKGFQGTT QTSH Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.
Terminal nucleotidyltransferase that catalyzes preferentially the transfert of ATP and GTP on RNA 3' poly(A) tail creating a heterogeneous 3' poly(A) tail leading to mRNAs stabilization by protecting mRNAs from active deadenylation (PubMed:21788334, PubMed:30026317). Also functions as a catalytic subunit of a TRAMP-like complex which has a poly(A) RNA polymerase activity and is involved in a post-transcriptional quality control mechanism. Polyadenylation with short oligo(A) tails is required for the degradative activity of the exosome on several of its nuclear RNA substrates. Doesn't need a cofactor for polyadenylation activity (in vitro) (PubMed:21788334, PubMed:21855801). Required for cytoplasmic polyadenylation of mRNAs involved in carbohydrate metabolism, including the glucose transporter SLC2A1/GLUT1 (PubMed:28383716). Plays a role in replication-dependent histone mRNA degradation, probably through terminal uridylation of mature histone mRNAs. May play a role in sister chromatid cohesion (PubMed:18172165). Mediates 3' adenylation of the microRNA MIR21 followed by its 3'-to-5' trimming by the exoribonuclease PARN leading to degradation (PubMed:25049417). Mediates 3' adenylation of H/ACA box snoRNAs (small nucleolar RNAs) followed by its 3'-to-5' trimming by the exoribonuclease PARN which enhances snoRNA stability and maturation (PubMed:22442037). [UniProt].
Specifications
| PAPD5 | |
| Polyclonal | |
| Unconjugated | |
| PAPD5 | |
| 5730445M16Rik; 5830428A09; GLD4; Non-canonical poly(A) RNA polymerase PAPD5; PAP associated domain containing 5; PAP-associated domain-containing protein 5; PAPD5; poly(A) RNA polymerase D5, non-canonical; Tent4b; Terminal guanylyltransferase; Terminal nucleotidyltransferase 4B; terminal uridylyltransferase 3; Topoisomerase-related function protein 4-2; TRF4-2; TUT3; TUTase 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64282 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NDF8 | |
| PAPD5 | |
| Recombinant protein corresponding to Human PAPD5. Recombinant protein control fragment (Product #RP-98641). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction