missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PAPD5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29369
This item is not returnable.
View return policy
Description
PAPD5 Polyclonal antibody specifically detects PAPD5 in Human samples. It is validated for Immunoprecipitation
Specifications
| PAPD5 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| EC 2.7.7, EC 2.7.7.-, FLJ40270, PAP associated domain containing 5, Terminal uridylyltransferase 3, Topoisomerase-related function protein 4-2, TRF4-2PAP-associated domain-containing protein 5, TUTase 3 | |
| Between 500 and 550 | |
| 100 μg | |
| Cancer | |
| 64282 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction