missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAPD4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £318.00
Specifications
| Antigen | PAPD4 |
|---|---|
| Dilution | Western Blot 1:1000, ELISA 1:100-1:1000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18356976
|
Novus Biologicals
NBP3-18647-25UL |
25 μg |
£139.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18328025
|
Novus Biologicals
NBP3-18647-100UL |
100 μg |
£318.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PAPD4 Polyclonal antibody specifically detects PAPD4 in Human samples. It is validated for Western Blot, ELISASpecifications
| PAPD4 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q6PIY7 | |
| 167153 | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000, ELISA 1:100-1:1000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Whole antiserum | |
| EC 2.7.7.19, FLJ38499, GLD2TUTase 2, hGLD-2, PAP associated domain containing 4, PAP-associated domain-containing protein 4, poly(A) RNA polymerase GLD2, Terminal uridylyltransferase 2 | |
| Polyclonal antibody raised in rabbit against human PAPD4 (PAP associated domain containing 4), using a KLH-conjugated synthetic peptide containing a sequence from the N-terminal part of the protein. | |
| Whole Antiserum |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title