missing translation for 'onlineSavingsMsg'
Learn More

Papaverine, TRC

Product Code. 30135424
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135424 500 mg 500mg
17921945 100 mg 100mg
17900595 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135424 Supplier Toronto Research Chemicals Supplier No. TRCP190503500MG
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-74-2
Molecular Formula C20 H21 N O4
Molecular Weight (g/mol) 339.38
Synonym Isoquinoline, 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-, Isoquinoline, 6,7-dimethoxy-1-veratryl- (8CI), 1-[(3,4-Dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline, 6,7-Dimethoxy-1-(3,4-dimethoxybenzyl)isoquinoline, 6,7-Dimethoxy-1-veratrylisoquinoline, NSC 136630, Papaverin, Papaverine
IUPAC Name 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline
SMILES COc1ccc(Cc2nccc3cc(OC)c(OC)cc23)cc1OC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C20H21NO4/c1-22-17-6-5-13(10-18(17)23-2)9-16-15-12-20(25-4)19(24-3)11-14(15)7-8-21-16/h5-8,10-12H,9H2,1-4H3
Formula Weight 339.1471
Percent Purity >95%
Chemical Name or Material Papaverine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.