missing translation for 'onlineSavingsMsg'
Learn More

Papaverine hydrochloride, 99%

Product Code. p-7028222
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427096 5 g 5g
11437096 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11427096 Supplier Thermo Scientific Alfa Aesar Supplier No. B25412.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 61-25-6 | C20H22ClNO4 | 375.849 g/mol

Papaverine hydrochloride is used as a smooth muscle relaxant, with noted treatment for impotence. It can also act as a cerebral vasodilator through the possible inhibition of PDE (phosphodiesterases) and/or calcium channel actions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (50 mg/ml), DMSO, methanol, 100% ethanol and chloroform.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-25-6
Molecular Formula C20H22ClNO4
Molecular Weight (g/mol) 375.849
MDL Number MFCD00012745
InChI Key UOTMYNBWXDUBNX-UHFFFAOYSA-N
Synonym papaverine hydrochloride, cardiospan, cardoverina, dispamil, drapavel, forpavin, papalease, papaversan, pavatest, paverolan
PubChem CID 6084
IUPAC Name 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline;hydrochloride
SMILES COC1=C(C=C(C=C1)CC2=NC=CC3=CC(=C(C=C32)OC)OC)OC.Cl

Specifications

Melting Point ∼220°C (decomposition)
Quantity 5 g
Merck Index 14,7019
Formula Weight 375.85
Percent Purity 99%
Chemical Name or Material Papaverine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.