missing translation for 'onlineSavingsMsg'
Få mere at vide

Papaverine hydrochloride, 99%

Artikelnummer. 11437096
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
5 g
25 g
Pakningsstørrelse:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
11437096 25 g 25g
11427096 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11437096 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. B25412.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 61-25-6 | C20H22ClNO4 | 375.849 g/mol

Papaverine hydrochloride is used as a smooth muscle relaxant, with noted treatment for impotence. It can also act as a cerebral vasodilator through the possible inhibition of PDE (phosphodiesterases) and/or calcium channel actions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (50 mg/ml), DMSO, methanol, 100% ethanol and chloroform.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 61-25-6
Molecular Formula C20H22ClNO4
Molecular Weight (g/mol) 375.849
MDL Number MFCD00012745
InChI Key UOTMYNBWXDUBNX-UHFFFAOYSA-N
Synonym papaverine hydrochloride, cardiospan, cardoverina, dispamil, drapavel, forpavin, papalease, papaversan, pavatest, paverolan
PubChem CID 6084
IUPAC Name 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline;hydrochloride
SMILES COC1=C(C=C(C=C1)CC2=NC=CC3=CC(=C(C=C32)OC)OC)OC.Cl

Tekniske data

Melting Point ∼220°C (decomposition)
Quantity 25 g
Merck Index 14,7019
Formula Weight 375.85
Percent Purity 99%
Chemical Name or Material Papaverine hydrochloride

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.