missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Papain (Papaya), 2X

Product Code. 16181780
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 16181780

Brand: Abnova™ P5245.25mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

Accession Number P00784
For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 23.4kDa
Name Papain (Papaya), 2x
Preparation Method Native protein purified from Papaya Latex
Quantity 25 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name PAPAIN
Assays Papain Assay; Cell isolation (more gentle than other proteases); Protein structural studies; Peptide mapping; Red cell surface modification for antibody screening or identification; Fab preparation from IgG and IgM antibodies; Solubilization of integral membrane proteins; Production of glycopeptides from purified proteoglycans; Enzymatic wound debridement.
Biological Activity Activates to at least 15 units per mg protein. One Unit hydrolyzes one micromole of benzoyl-L-arginine ethyl ester per minute at 25°C, pH 6.2, after activation in a solution containing 1.1 mM EDTA, 0.067 mM mercaptoethanol and 5.5 mM cysteine-HCl for 30 minutes.
Species Plants
Protein Tag None
Expression System Native protein purified from Papaya Latex
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.