missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAOX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18972-1-AP-150UL
This item is not returnable.
View return policy
Description
Flavoenzyme which catalyzes the oxidation of N(1)-acetylspermine to spermidine and is thus involved in the polyamine back-conversion. Can also oxidize N(1)-acetylspermidine to putrescine. Substrate specificity: N(1)-acetylspermine = N(1)-acetylspermidine > N(1),N(12)-diacylspermine >> spermine. Does not oxidize spermidine. Plays an important role in the regulation of polyamine intracellular concentration and has the potential to act as a determit of cellular sensitivity to the antitumor polyamine analogs.Specifications
| PAOX | |
| Polyclonal | |
| Unconjugated | |
| PAOX | |
| PAO, PAOX, Polyamine oxidase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 196743, 212503, 293589 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6QHF9, Q8C0L6 | |
| PAOX | |
| PAOX Fusion Protein Ag5777 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction