missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAOX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525758
This item is not returnable.
View return policy
Description
PAOX Polyclonal Antibody for Western Blot, IHC (P)
Flavoenzyme which catalyzes the oxidation of N(1)-acetylspermine to spermidine and is thus involved in the polyamine back-conversion (PubMed:12477380). Can also oxidize N(1)-acetylspermidine to putrescine. Substrate specificity: N(1)-acetylspermine = N(1)-acetylspermidine > N(1),N(12)-diacylspermine >> spermine. Does not oxidize spermidine. Plays an important role in the regulation of polyamine intracellular concentration and has the potential to act as a determinant of cellular sensitivity to the antitumor polyamine analogs (PubMed:12477380). [UniProt].
Specifications
| PAOX | |
| Polyclonal | |
| Unconjugated | |
| Paox | |
| 2410012F02Rik; AI118225; exo-N4-amino; PAO; Paox; Peroxisomal N(1)-acetyl-spermine/spermidine oxidase; peroxisomal N1-acetyl-spermine/spermidine oxidase; polyamine oxidase; polyamine oxidase (exo-N4-amino); polyamine oxidase (exo-N4-amino) isoform 1; putative GTP-binding protein; UNQ1923/PRO4398 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 196743, 212503 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6QHF9, Q8C0L6 | |
| Paox | |
| KLH conjugated synthetic peptide between 467-496 amino acids from the central region of human PAOX. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction