missing translation for 'onlineSavingsMsg'
Learn More

Pantoprazole Sulfone, TRC

Product Code. 30006943
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006943 100 mg 100mg
30004366 25 mg 25mg
30004963 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006943 Supplier Toronto Research Chemicals Supplier No. TRCP183016100MG
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127780-16-9
Molecular Formula C16 H15 F2 N3 O5 S
Molecular Weight (g/mol) 399.37
Synonym 5-(Difluoromethoxy)-2-[[(3,4-dimethoxypyridin-2-yl)methyl]sulphonyl]-1H-benzimidazole, Pantoprazole Sodium Sesquihydrate Imp. A (EP), Pantoprazole Sulphone
IUPAC Name 5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methylsulfonyl]-1H-benzimidazole
SMILES COc1ccnc(CS(=O)(=O)c2nc3cc(OC(F)F)ccc3[nH]2)c1OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H15F2N3O5S/c1-24-13-5-6-19-12(14(13)25-2)8-27(22,23)16-20-10-4-3-9(26-15(17)18)7-11(10)21-16/h3-7,15H,8H2,1-2H3,(H,20,21)
Formula Weight 399.07
Percent Purity >95%
Chemical Name or Material Pantoprazole Sulfone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.