missing translation for 'onlineSavingsMsg'
Learn More

Pantoprazole Sulfide, TRC

Product Code. 30011702
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011702 25 mg 25mg
30003292 250 mg 250mg
17984195 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011702 Supplier Toronto Research Chemicals Supplier No. TRCP18302025MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102625-64-9
Molecular Formula C16 H15 F2 N3 O3 S
Molecular Weight (g/mol) 367.37
Synonym 5-(Difluoromethoxy)-2-[[(3,4-dimethoxypyridin-2-yl)methyl]sulphanyl]-1H-benzimidazole, Pantoprazole Sodium Sesquihydrate Imp. B (EP)
IUPAC Name 5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methylsulfanyl]-1H-benzimidazole
SMILES COc1ccnc(CSc2nc3cc(OC(F)F)ccc3[nH]2)c1OC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H15F2N3O3S/c1-22-13-5-6-19-12(14(13)23-2)8-25-16-20-10-4-3-9(24-15(17)18)7-11(10)21-16/h3-7,15H,8H2,1-2H3,(H,20,21)
Formula Weight 367.0802
Percent Purity >95%
Chemical Name or Material 5-(Difluoromethoxy)-2-([(3,4-dimethoxypyridin-2-yl)methyl]sulphanyl)-1H-benzimidazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.