missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pantethine, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Pantethine is a dimeric form of pantothenic acid, is an intermediate in the production of Coenzyme A, is available as a dietary supplement, and is used to treat acne and improve the blood-cholesterol profile.
Specifications
Specifications
| Chemical Name or Material | Pantethine |
| Physical Form | Crystals |
| CAS | 16816-67-4 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H42N4O8S2 |
| Synonym | D-Pantethine LBF disulfide |
| Solubility Information | H2O : ≥ 100 mg/mL (180.27 mM) ∣DMSO : ≥ 100 mg/mL (180.27 mM) ∣Ethanol : 100 mg/mL (180.27 mM; Need ultrasonic) |
| SMILES | O=C(NCCSSCCNC(CCNC([C@H](O)C(C)(C)CO)=O)=O)CCNC([C@H](O)C(C)(C)CO)=O |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction