missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PANK2 Antibody (OTI3H9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73241CL1
This item is not returnable.
View return policy
Description
PANK2 Monoclonal antibody specifically detects PANK2 in Canine,Human,Monkey,Mouse,Primate,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| PANK2 | |
| Monoclonal | |
| CoraFluor 1 | |
| C20orf48, EC 2.7.1.33, FLJ17232, Hallervorden-Spatz syndrome, HARPFLJ11729, hPanK2, HSS, MGC15053, NBIA1, pantothenate kinase 2, pantothenate kinase 2, mitochondrial, Pantothenic acid kinase 2, PKANneurodegeneration with brain iron accumulation 1 (Hallervorden-Spatz syndrome) | |
| Full length human recombinant protein of human PANK2(NP_705902) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Canine, Human, Monkey, Mouse, Primate | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI3H9 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 80025 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction