missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PANK1/2/3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12466-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein belonging to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. This family member is expressed most abundantly in the liver.Specifications
| PANK1/2/3 | |
| Polyclonal | |
| Unconjugated | |
| PANK1 | |
| hPanK3, PANK1/2/3, PANK3, pantothenate kinase 3, Pantothenic acid kinase 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 211347, 294088, 296167, 360511, 53354, 74450, 75735, 79646, 80025 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8K4K6, Q8R2W9, Q8TE04, Q9BZ23, Q9H999 | |
| Pank1, Pank2, Pank3 | |
| PANK1/2/3 Fusion Protein Ag3140 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction