missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Panendothelial Cell Antigen Antibody (MECA-32), mFluor Violet 450 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-77668MFV450
This item is not returnable.
View return policy
Description
Panendothelial Cell Antigen Monoclonal antibody specifically detects Panendothelial Cell Antigen in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
| Panendothelial Cell Antigen | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| MECA-32 | |
| Flow Cytometry | |
| FELS, Fenestrated endothelial-linked structure protein, gp68, MECA32, MECA-32, Panendothelial Cell Antigen, plasmalemma vesicle associated protein, Plasmalemma Vesicle Protein 1, PV-1 protein | |
| Mouse lymph node stromal cells. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 84094 | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction