Learn More
Applied Critical Fluids Panel-Mount Coupling Insert for 1/4-28 Port, PP
Description
- Couplings rotate in socket to eliminate tubing kinks without loosening compression nuts.
- Useful in a wide variety of lab applications such as spectrometry, chromatography, and electrophoresis.
- Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
- Plug insert into body to connect fitting—fitting halves lock into place immediately.
- To disconnect fitting halves, simply press the thumb latch on the coupling body.
- These fittings simplify disconnecting and reconnecting your fluid lines.
- The 1/4-28 flat-bottom tubing ports with compression nuts provide a secure connection to eliminate worries of tubing line leak at the fitting.
- The audible click when the coupling engages confirms a solid connection.
- The couplings feature an integral shutoff valve; when the coupling disconnects, the valve stops flow to keep your process uncontaminated.
- Once installed, the couplings can save lab technicians valuable time by eliminating the need to thread/rethread the tubing line when replacing a fitting.
Specifications
Specifications
| Material | Polypropylene |
| Tubing Size | 1/8 in. |
| Product Type | Quick-disconnect x UNF(F) |