missing translation for 'onlineSavingsMsg'
Learn More

Pancreatic trypsin inhibitor Antibody (35), DyLight 405, Novus Biologicals™

Product Code. 18422588 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422588 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422588 Supplier Novus Biologicals Supplier No. NBP223590V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Pancreatic trypsin inhibitor Monoclonal antibody specifically detects Pancreatic trypsin inhibitor in Bovine samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pancreatic trypsin inhibitor
Applications Western Blot, ELISA
Classification Monoclonal
Clone 35
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias Aprotinin, Basic protease inhibitor, BPI, BPTI, PTI, PTI pancreatic trypsin inhibitor
Host Species Mouse
Immunogen Aprotinin from bovine lung
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 404172
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.