missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Pancreatic trypsin inhibitor Monoclonal antibody specifically detects Pancreatic trypsin inhibitor in Bovine samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Pancreatic trypsin inhibitor |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 18 |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | Aprotinin, Basic protease inhibitor, BPI, BPTI, PTI, PTI pancreatic trypsin inhibitor |
| Host Species | Mouse |
| Immunogen | Aprotinin from bovine lung. |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?