missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic Lipase Rabbit anti-Human, Clone: 120, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90231-50UL
This item is not returnable.
View return policy
Description
Pancreatic Lipase Monoclonal specifically detects Pancreatic Lipase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Pancreatic Lipase | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 5406 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 120 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| EC 3.1.1.21, EC 3.1.1.3, pancreatic lipasepancreatic triacylglycerol lipase, PL, PTL, triacylglycerol acylhydrolase | |
| Recombinant Human Pancreatic Lipase Protein | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction