missing translation for 'onlineSavingsMsg'
Learn More

Pancreatic Lipase Antibody (PNLIP/9040) [Janelia Fluor™ 669], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP3-26989JF669

Product Code. 30160197

  • £491.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Pancreatic Lipase Monoclonal antibody specifically detects Pancreatic Lipase in Human samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pancreatic Lipase
Monoclonal
Janelia Fluor 669
EC 3.1.1.21, EC 3.1.1.3, pancreatic lipasepancreatic triacylglycerol lipase, PL, PTL, triacylglycerol acylhydrolase
Recombinant fragment (around aa 265-465) of human Pancreatic Lipase protein (exact sequence is proprietary)
0.1 mL
Primary
Human
Purified
ELISA, Immunohistochemistry (Paraffin)
PNLIP/9040
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
5406
Store at 4°C in the dark.
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.