missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Pancreatic Lipase Antibody (PNLIP/8913), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-26711-100ug

 View more versions of this product

Product Code. 30043843

  • £351.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Pancreatic Lipase Monoclonal antibody specifically detects Pancreatic Lipase in Human samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pancreatic Lipase
Monoclonal
0.2 mg/mL
ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
EC 3.1.1.21, EC 3.1.1.3, pancreatic lipasepancreatic triacylglycerol lipase, PL, PTL, triacylglycerol acylhydrolase
Recombinant fragment (around aa 1-200) of human Pancreatic Lipase protein (exact sequence is proprietary)
100 μg
Primary
Human
Purified
ELISA, Immunohistochemistry (Paraffin)
PNLIP/8913
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
5406
Store at 4°C.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.