missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pancreatic Amylase Monoclonal Antibody (501)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA134918
This item is not returnable.
View return policy
Description
MA1-34918 detects Pancreatic Amylase from human samples.
Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch andglycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas.
Specifications
| Pancreatic Amylase | |
| Monoclonal | |
| 0.6 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P04746, P19961 | |
| AMY2A, AMY2B | |
| Human Pancreatic Amylase. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
| ELISA, Immunohistochemistry (Frozen), Immunoprecipitation, Neutralization, Immunocytochemistry | |
| 501 | |
| Unconjugated | |
| AMY2A | |
| 1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; alpha-amylase; Alpha-amylase 2B; Amy2; Amy2-1; Amy2-2; Amy2a; Amy2a1; Amy2a4; AMY2B; AMY3; amylase; amylase 2-2, pancreatic; amylase 2a1; amylase 2a4; amylase 2a4, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2A (pancreatic); amylase, alpha 2B (pancreatic); Amy-X; carcinoid alpha-amylase; ENSMUSG00000074267; glycogenase; HXA; mAmy2-1; mAmy2-3; mAmy2-6; OTTMUSG00000022432; OTTMUSG00000022433; OTTMUSG00000022462; PA; Pancreatic alpha-amylase; pancreatic amylase alpha 2A | |
| Mouse | |
| PEG precipitation | |
| RUO | |
| 279, 280 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction