missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic Amylase Alpha Antibody (OTI6D4), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73238PCP
This item is not returnable.
View return policy
Description
Pancreatic Amylase Alpha Monoclonal antibody specifically detects Pancreatic Amylase Alpha in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Pancreatic Amylase Alpha | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 279 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI6D4 | |
| Western Blot, Immunohistochemistry | |
| AMY2, AMY2A amylase, alpha 2A (pancreatic), AMY2B, PA, pancreatic alpha-amylase | |
| Recombinant protein expressed in E.coli corresponding to amino acids 300-511 of human AMY2A | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction