missing translation for 'onlineSavingsMsg'
Learn More

Pancreatic Amylase Alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£164.00 - £442.00

Specifications

Antigen Pancreatic Amylase Alpha
Dilution Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30231914
View Documents
Novus Biologicals
NBP3-38499-20ul
20 μL
£164.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30228848
View Documents
Novus Biologicals
NBP3-38499-100ul
100 μL
£442.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pancreatic Amylase Alpha Polyclonal antibody specifically detects Pancreatic Amylase Alpha in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications

Specifications

Pancreatic Amylase Alpha
ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
Rabbit
Human, Mouse, Rat
AMY2, AMY2A amylase, alpha 2A (pancreatic), AMY2B, PA, pancreatic alpha-amylase
Recombinant fusion protein containing a sequence corresponding to amino acids 16-511 of human Pancreatic Amylase Alpha (NP_000690.1).,, Sequence:, KPFIYQEVIDLGGEPIKSSDYFGNGRVTEFKYGAKLGTVIRKWNGEKMSYLKNWGEGWGFVPSDRALVFVDNHDNQRGHGAGGASILTFWDARLYKMAVGFMLAHPYGFTRVMSSYRWPRQFQNGNDVNDWVGPPNNNGVIKEVTINPDTTCGNDWVCEHRWRQIRNMVIFRNVVDGQPFTNWYDNGSNQVAFGRGNRGFIVFNNDDWSFSLTLQTGLPAGTYCDVISGDKINGNCTGIKIYVSDDGKAHFSISNSAEDPFIAIHAESKL
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
279
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.