missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Panaxadiol

Product Code. 16479028
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479028 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479028 Supplier MedChemExpress Supplier No. HYN0596

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Panaxadiol is found in the roots of Panax ginseng, activities neuroprotective and anti-tumour functions. Panaxadiol inhibits programmed cell death-ligand 1(PD-L1) expression and tumour proliferation.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 460.73 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 19666-76-3
Product Type Natural Products
Purity 98.0%
Solubility DMSO : 100 mg/mL (217.05 mM∣Need ultrasonic) ∣Ethanol : 20 mg/mL (43.41 mM∣Need ultrasonic)
Synonym 20(R)-Panaxadiol
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.