missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAN3 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44286CL1
This item is not returnable.
View return policy
Description
PAN3 Polyclonal antibody specifically detects PAN3 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| PAN3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| hPan3, PAB-dependent poly(A)-specific ribonuclease subunit 3, PABP1-dependent poly A-specific ribonuclease subunit PAN3, PABP-dependent poly(A) nuclease 3, PAN3 poly(A) specific ribonuclease subunit homolog (S. cerevisiae), TS | |
| The epitope recognized by this antibody maps to a region between residue 175 to 225 of human PAB-dependent poly using the numbering given in entry Q58A45.3 (GeneID 255967). | |
| 0.1 mL | |
| Protein Kinase | |
| 255967 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction