missing translation for 'onlineSavingsMsg'
Learn More

PAN Indicator, 0.1% (w/v) in Denatured Alcohol, Indicator for Complexometric Titrations, Ricca

Product Code. 31940531
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31940531 100 mL 100mL
31940709 1 L 1 L
31940750 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31940531 Supplier Ricca Chemical Company Supplier No. R5487000100C
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • PAN Indicator
  • Available in 100 mL, 500 mL, and 1 L package sizes in amber glass containers

Specifications

CAS 64-17-5, 67-56-1, 85-85-8
Packaging Amber Glass Container
Color Amber
Quantity 100 mL
Grade ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.