missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pan Coronavirus Spike Antibody (39), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14661DL594
This item is not returnable.
View return policy
Description
Pan Coronavirus Spike Monoclonal antibody specifically detects Pan Coronavirus Spike in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Pan Coronavirus Spike | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| This product is a recombinant monoclonal antibody expressed from HEK293 cells. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| 39 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction