missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pan Coronavirus Spike Antibody (38), Janelia Fluor™ 646, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Pan Coronavirus Spike Monoclonal antibody specifically detects Pan Coronavirus Spike in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | Pan Coronavirus Spike |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 38 |
| Conjugate | Janelia Fluor 646 |
| Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | This product is a recombinant monoclonal antibody expressed from HEK293 cells. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?