missing translation for 'onlineSavingsMsg'
Learn More

Pan Coronavirus Spike Antibody (38), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18349471
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349471 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349471 Supplier Bio-Techne Supplier No. NBP314660AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Pan Coronavirus Spike Monoclonal antibody specifically detects Pan Coronavirus Spike in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pan Coronavirus Spike
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 38
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen This product is a recombinant monoclonal antibody expressed from HEK293 cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.