missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pan Cadherin Antibody (0J8Q6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£159.00 - £365.00
Specifications
| Antigen | pan Cadherin |
|---|---|
| Clone | 0J8Q6 |
| Dilution | Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18623057
|
Novus Biologicals
NBP3-05342-100ul |
100 μg |
£365.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18666027
|
Novus Biologicals
NBP3-05342-20ul |
20 μg |
£159.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
pan Cadherin Monoclonal antibody specifically detects pan Cadherin in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| pan Cadherin | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 373-622 of human pan Cadherin (P12830). PIFNPTTYKGQVPENEANVVITTLKVTDADAPNTPAWEAVYTILNDDGGQFVVTTNPVNNDGILKTAKGLDFEAKQQYILHVAVTNVVPFEVSLTTSTATVTVDVLDVNEAPIFVPPEKRVEVSEDFGVGQEITSYTAQEPDTFMEQKITYRIWRDTANWLEINPDTGAISTRAELDREDFEHVKNSTYTALIIATDNGSPVATGTGTLLLILSDVNDNAPIPEPRTIFFCERNPKPQVINIIDADLPPN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 0J8Q6 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 999 | |
| IgG2b κ | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title