missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAMR1 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149215
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
PAMR1 may be involved in regeneration of skeletal muscle. Although related to peptidase S1 family, lacks the conserved active Ser residue in position 665 which is replaced by a Thr, suggesting that it has no protease activity.
Specifications
| PAMR1 | |
| Polyclonal | |
| Unconjugated | |
| PAMR1 | |
| 5930437L24; DKFZP586H2123; E430002G05Rik; FP938; inactive serine protease PAMR1; Pamr1; peptidase domain containing associated with muscle regeneration 1; peptidase domain-containing protein associated with muscle regeneration 1; Ramp; regeneration associated muscle protease; regeneration-associated muscle protease homolog; RGD1308745; UNQ699/PRO1344 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 25891 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q6UXH9 | |
| PAMR1 | |
| Mouse myeloma cell line NS0-derived recombinant human PAMR1, Leu22-Lys737. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction