missing translation for 'onlineSavingsMsg'
Learn More

PAMR1 Antibody [Biotin], Novus Biologicals™

Product Code. 30734434 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734434 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734434 Supplier Novus Biologicals Supplier No. AF6517B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

PAMR1 Polyclonal antibody specifically detects PAMR1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAMR1
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias DKFZP586H2123, peptidase domain containing associated with muscle regeneration 1, Peptidase domain-containing protein associated with muscle regeneration 1, RAMPinactive serine protease PAMR1, regeneration associated muscle protease, Regeneration-associated muscle protease homolog
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human PAMR1, Leu22-Lys737, Accession # NP_056245
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25891
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.