missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pamoic Acid, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Pamoic Acid
Specifications
Specifications
| Chemical Name or Material | Embonic Acid |
| CAS | 130-85-8 |
| Analyte or Component Names | Embonic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H16 O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4,4'-Methylenebis[3-hydroxy-2-naphthalenecarboxylic acid], 2-Naphthoic acid, 4,4'-methylenebis[3-hydroxy- (6CI,7CI,8CI), 4,4'-Methylenebis[3-hydroxy-2-naphthalenecarboxylic acid], 2,2'-Dihydroxy-1,1'-dinaphthylmethane-3,3'-dicarboxylic acid, 4,4'-Methylenebis(3-hydroxy-2-naphthalenecarboxylic acid), 4,4'-Methylenebis[3-hydroxy-2-naphthoic acid], 4-[(3-Carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid, Bis(2-hydroxy-3-carboxy-1-naphthyl)methane, Embonic acid, KG 122, NSC 30188, NSC 40132, Pamoic Acid |
| InChI Formula | InChI=1S/C23H16O6/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29/h1-10,24-25H,11H2,(H,26,27)(H,28,29) |
| SMILES | OC(=O)c1cc2ccccc2c(Cc3c(O)c(cc4ccccc34)C(=O)O)c1O |
| IUPAC Name | 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction