missing translation for 'onlineSavingsMsg'
Learn More

Pamidronate disodium pentahydrate, MedChemExpress

Product Code. 16488588
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488588 10 mg 10mg
16404467 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488588 Supplier Medchem Express Supplier No. HYB0730

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pamidronate disodium pentahydrate is a nitrogen-containing bisphosphonate, used to prevent osteoporosis.

Pamidronate disodium pentahydrate is a nitrogen-containing bisphosphonate, used to prevent osteoporosis.

Specifications

Chemical Name or Material Pamidronate disodium pentahydrate
Physical Form Solid
CAS 109552-15-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C3H19NNa2O12P2
Solubility Information H2O : 7.14 mg/mL (19.34 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.71 mM; ultrasonic and warming and heat to 80°C)
SMILES OC(P(O)(O[Na])=O)(CCN)P(O)(O[Na])=O.O.O.O.O.O
Molecular Weight (g/mol) 369.11
Formula Weight 369.11
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.