missing translation for 'onlineSavingsMsg'
Learn More

Pamicogrel, MedChemExpress

Product Code. 16482418
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482418 5 mg 5mg
16460219 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482418 Supplier Medchem Express Supplier No. HYU00175
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pamicogrel (KBT3022) is a cyclooxygenase (COX) inhibitor.

Pamicogrel (KBT3022) is a cyclooxygenase (COX) inhibitor.

Specifications

Chemical Name or Material Pamicogrel
CAS 101001-34-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H24N2O4S
Quantity 5 mg
Synonym KBT3022
Solubility Information DMSO : 83.33 mg/mL (185.78 mM; Need ultrasonic)
SMILES O=C(OCC)CN1C(C2=NC(C3=CC=C(OC)C=C3)=C(C4=CC=C(OC)C=C4)S2)=CC=C1
Molecular Weight (g/mol) 448.53
Formula Weight 448.53
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.