missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAMCI Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£231.00 - £513.00
Specifications
| Antigen | PAMCI |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18459540
|
Novus Biologicals
NBP1-80937-25ul |
25ul |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18218975
|
Novus Biologicals
NBP1-80937 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PAMCI Polyclonal specifically detects PAMCI in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PAMCI | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| PAM COOH-terminal interactor protein 1, PAMCI, PCIP1, P-CIP1peptidylglycine alpha-amidating monooxygenase COOH-terminal interactorprotein-1, Peptidylglycine alpha-amidating monooxygenase COOH-terminal interactor, Ras association (RalGDS/AF-6) domain family (N-terminal) member 9, ras association domain-containing protein 9 | |
| RASSF9 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Mouse, Human | |
| O75901 | |
| 9182 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:YRILIDKLSAEIEKEVKSVCIDINEDAEGEAASELESSNLESVKCDLEKSMKAGLKIHSHLSGIQKEIKYSDSLLQMKAKEY | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title