missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567939
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
SNX33 (sorting nexin-33), also known as SH3PX3, SH3PXD3C or SNX30, is a 574 amino acid protein that interacts with ADAM15 and FAS-L. Belonging to the sorting nexin family, SNX33 contains one BAR domain, one PX (phox homology) domain and one SH3 domain. The gene that encodes SNX33 consists of over 14,000 bases and maps to human chromosome 15q24.2. Housing approximately 106 million base pairs and encoding more than 700 genes, chromosome 15 makes up about 3% of the human genome. Angelman and Prader-Willi syndromes are associated with loss of function or deletion of genes in the 15q11-q13 region.
Specifications
| PAM | |
| Polyclonal | |
| Unconjugated | |
| PAM | |
| AU023734; AW107953; AW546647; C130061D10Rik; E3 ubiquitin-protein ligase MYCBP2; Highwire; KIAA0916; MYC binding protein 2; MYC binding protein 2, E3 ubiquitin protein ligase; Myc-binding protein 2; Mycbp2; Myc-bp2; PAL; PAM; pam, highwire, rpm 1; pam/highwire/rpm-1 protein; pancreatic peptidylglycine alpha-amidating monooxygenase; peptidyl alpha-amidating enzyme; peptidyl-alpha-hydroxyglycine alpha-amidating lyase; peptidylamidoglycolate lyase; peptidylglycine 2-hydroxylase; Peptidyl-glycine alpha-amidating monooxygenase; peptidylglycine alpha-amidating monooxygenase; PEPTIDYL-GLYCINE ALPHA-AMIDATING MONOOXYGENASE PRECURSOR (PAM); Peptidylglycine alpha-hydroxylating monooxygenase; PHM; Phr; Phr1; probable E3 ubiquitin-protein ligase MYCBP2; protein associated with Myc; Protein Magellan; R75243 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18484, 25508, 5066 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P14925, P19021, P97467 | |
| PAM | |
| Synthesized peptide derived from human PAM. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction