missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAM Monoclonal Antibody (K49041_3E6), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | PAM |
|---|---|
| Clone | K49041_3E6 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30205805
|
Invitrogen™
MA555849 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30205433
|
Invitrogen™
MA556229 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: VKSDMVMMHE HHKETEYKDK IPLLQQPKRE EEEVLDQGDF YSLLSKLLGE REDVVHVHKY NPTEKAESES DLVAEIANVV QKKDLGRSDA REGAEHERGN
PAM Monoclonal antibody specifically detects PAM in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| PAM | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AU023734; AW107953; AW546647; C130061D10Rik; E3 ubiquitin-protein ligase MYCBP2; Highwire; KIAA0916; MYC binding protein 2; MYC binding protein 2, E3 ubiquitin protein ligase; Myc-binding protein 2; Mycbp2; Myc-bp2; PAL; PAM; pam, highwire, rpm 1; pam/highwire/rpm-1 protein; pancreatic peptidylglycine alpha-amidating monooxygenase; peptidyl alpha-amidating enzyme; peptidyl-alpha-hydroxyglycine alpha-amidating lyase; peptidylamidoglycolate lyase; peptidylglycine 2-hydroxylase; Peptidyl-glycine alpha-amidating monooxygenase; peptidylglycine alpha-amidating monooxygenase; PEPTIDYL-GLYCINE ALPHA-AMIDATING MONOOXYGENASE PRECURSOR (PAM); Peptidylglycine alpha-hydroxylating monooxygenase; PHM; Phr; Phr1; probable E3 ubiquitin-protein ligase MYCBP2; protein associated with Myc; Protein Magellan; R75243 | |
| Recombinant human PAM (immunogen range is 351-450). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| PAM |
| K49041_3E6 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P19021 | |
| 5066 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title