missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PALS1/MPP5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29016
This item is not returnable.
View return policy
Description
PALS1/MPP5 Polyclonal antibody specifically detects PALS1/MPP5 in Human samples. It is validated for Immunoprecipitation
Specifications
| PALS1/MPP5 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 64398 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| MAGUK p55 subfamily member 5, membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5), PALS1FLJ12615, stardust | |
| Between 225 and 275 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction