missing translation for 'onlineSavingsMsg'
Learn More

Palovarotene, MedChemExpress

Product Code. 16492307
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492307 50 mg 50mg
16428777 5 mg 5mg
16426178 10 mg 10mg
16416556 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16492307 Supplier Medchem Express Supplier No. HY14799
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Palovarotene is a nuclear retinoic acid receptor γ (RAR-γ) agonist.

Palovarotene is a nuclear retinoic acid receptor γ (RAR-γ) agonist.

Specifications

Chemical Name or Material Palovarotene
CAS 410528-02-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H30N2O2
Quantity 50 mg
Synonym R 667 Ro 3300074
Solubility Information DMSO : 25 mg/mL (60.31 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C(/C=C/C2=C(CN3N=CC=C3)C=C4C(C)(C)CCC(C)(C)C4=C2)C=C1
Molecular Weight (g/mol) 414.54
Formula Weight 414.54
Percent Purity 95.9%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.