missing translation for 'onlineSavingsMsg'
Learn More

Palonosetron hydrochloride, MedChemExpress

Product Code. 16424157
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16424157 50 mg 50mg
16411356 200 mg 200mg
16407996 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16424157 Supplier Medchem Express Supplier No. HYA0021
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Palonosetron hydrochloride is a 5-HT3 antagonist used in the prevention and treatment of chemotherapy-induced nausea and vomiting (CINV).

Palonosetron hydrochloride is a 5-HT3 antagonist used in the prevention and treatment of chemotherapy-induced nausea and vomiting (CINV).

Specifications

Chemical Name or Material Palonosetron hydrochloride
CAS 135729-62-3
Color White
Purity Grade Notes Research
Molecular Formula C19H25ClN2O
Quantity 50 mg
Solubility Information H2O : 100 mg/mL (300.42 mM; Need ultrasonic) ∣DMSO : 3.33 mg/mL (10.00 mM; Need ultrasonic)
SMILES O=C1N(C[C@@]([H])(CCC2)C3=C2C=CC=C13)[C@@H]4CN5CCC4CC5.[H]Cl
Molecular Weight (g/mol) 332.87
Formula Weight 332.87
Percent Purity 99.58%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.