missing translation for 'onlineSavingsMsg'
Learn More

Palomid 529, MedChemExpress

Product Code. 16466496
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16466496 100 mg 100mg
16436108 10 mg 10mg
16418707 5 mg 5mg
16412247 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16466496 Supplier Medchem Express Supplier No. HY14581
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Palomid 529 is a potent inhibitor of mTORC1 and mTORC2 complexes.

Palomid 529 is a potent inhibitor of mTORC1 and mTORC2 complexes.

Specifications

Chemical Name or Material Palomid 529
CAS 914913-88-5
Color White
Purity Grade Notes Research
Molecular Formula C24H22O6
Quantity 100 mg
Synonym P529
Solubility Information DMSO : 100 mg/mL (246.04 mM; Need ultrasonic)
SMILES COC1=C(OCC2=CC=C(C=C2)OC)C=C3C(C4=C(C(O3)=O)C=C(C(C)O)C=C4)=C1
Molecular Weight (g/mol) 406.43
Formula Weight 406.43
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.