missing translation for 'onlineSavingsMsg'
Learn More

Palmitoylethanolamide, TRC

Product Code. 30005112
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005112 10 mg 10mg
30004167 50 mg 50mg
30010519 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005112 Supplier Toronto Research Chemicals Supplier No. TRCP15508010MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-31-0
Molecular Formula C18H37NO2
Molecular Weight (g/mol) 299.49
Synonym (Hydroxyethyl)palmitamide, 2-(Palmitoylamino)ethanol, 2-Palmitamidoethanol, AM 3112, Impulsin, Loramine P 256, Mackpeart DR 14V, N-(2-Hydroxyethyl)hexadecanamide, N-(2-Hydroxyethyl)palmitamide, N-Hexadecanoylethanolamine, N-Palmitoylethanolamine, NSC 23320, Palmidrol, Palmitic Acid Monoethanolamide, Palmitic Monoethanolamide
IUPAC Name N-(2-hydroxyethyl)hexadecanamide
SMILES CCCCCCCCCCCCCCCC(=O)NCCO

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H37NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)19-16-17-20/h20H,2-17H2,1H3,(H,19,21)
Formula Weight 299.2824
Percent Purity >95%
Chemical Name or Material Palmitoyl Ethanolamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.