missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Palmitoylethanolamide
GREENER_CHOICE

Product Code. 18095762
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18095762 50 mg 50mg
18054952 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18095762 Supplier Tocris Bioscience™ Supplier No. 0879/50
  • ONLINE EXCLUSIVE PRICE
    £380.00
    £359.10 / 50mg
    Save £20.90
    5% Off
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective GPR55 agonist. FAAH and PAA substrate

Endogenous lipid that acts as a selective GPR55 agonist (EC50 values are 4, 19 800 and > 30 000 nM at GPR55, CB2 and CB1 receptors respectively). Substrate for fatty acid amide hydrolase (FAAH) and PEA-preferring acid amidase (PAA) and exhibits antinociceptive and anticonvulsant in vivo. Directly activates PPARα (EC50 = 3 μM) producing robust anti-inflammatory actions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-31-0
Molecular Formula C18H37NO2

Specifications

Quantity 50 mg
Formula Weight 299.5
Purity >98%
Molecular Weight (g/mol) 299.499
Chemical Name or Material Palmitoylethanolamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.