missing translation for 'onlineSavingsMsg'
Learn More

Palmitoylethanolamide, MedChemExpress

Product Code. 16468018
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468018 10 mg 10mg
16427796 100 mg 100mg
16403947 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16468018 Supplier Medchem Express Supplier No. HY20685
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Palmitoylethanolamide (Palmidrol) is an active endogenous compound which can used for preventing virus infection of the respiratory tract.

Palmitoylethanolamide (Palmidrol) is an active endogenous compound which can used for preventing virus infection of the respiratory tract.

Specifications

Chemical Name or Material Palmitoylethanolamide
Physical Form Powder
CAS 544-31-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H37NO2
For Use With (Application) COVID-19-anti-virus
Synonym PalmidrolLoramine P 256
Solubility Information DMSO : 2.6 mg/mL (8.68 mM; Need ultrasonic)
SMILES CCCCCCCCCCCCCCCC(NCCO)=O
Molecular Weight (g/mol) 299.49
Formula Weight 299.49
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.