missing translation for 'onlineSavingsMsg'
Learn More

Palmitoylethanolamide, MedChemExpress

Product Code. 16427796
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16427796

Brand: Medchem Express HY20685

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Palmitoylethanolamide (Palmidrol) is an active endogenous compound which can used for preventing virus infection of the respiratory tract.

Palmitoylethanolamide (Palmidrol) is an active endogenous compound which can used for preventing virus infection of the respiratory tract.

Specifications

Chemical Name or Material Palmitoylethanolamide
Physical Form Powder
CAS 544-31-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H37NO2
For Use With (Application) COVID-19-anti-virus
Synonym PalmidrolLoramine P 256
Solubility Information DMSO : 2.6 mg/mL (8.68 mM; Need ultrasonic)
SMILES CCCCCCCCCCCCCCCC(NCCO)=O
Molecular Weight (g/mol) 299.49
Formula Weight 299.49
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.