missing translation for 'onlineSavingsMsg'
Learn More

Palmitoyl Chloride, TRC

Product Code. 30420849
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420849 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420849 Supplier Toronto Research Chemicals Supplier No. TRCP1564801G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Palmitoyl Chloride
CAS 112-67-4
Purity Grade Notes GC
Molecular Formula C16H31ClO
Quantity 1 g
Synonym Hexadecanoic Acid Chloride, NSC 9854, Palmitic Acid Chloride, Palmitic Chloride, Palmityl Acid Chloride
InChI Formula InChI=1S/C16H31ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3
SMILES CCCCCCCCCCCCCCCC(=O)Cl
IUPAC Name hexadecanoyl chloride
Molecular Weight (g/mol) 274.87
Formula Weight 274.2063
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.