Learn More
Palmitoyl chloride, 98%, ACROS Organics™
Brand: Acros Organics 304295000
Code : 29
Additional Details : CAS Number : 112-67-4 Weight : 0.50250kg Transport : UN number : 3265 Chem class : 8 Pack group : II
Packaging | Glass bottle |
---|---|
Quantity | 500mL |
Chemical Identifiers
112-67-4 | |
274.88 | |
ARBOVOVUTSQWSS-UHFFFAOYSA-N | |
8206 | |
CCCCCCCCCCCCCCCC(=O)Cl |
C16H31ClO | |
MFCD00000742 | |
palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride | |
hexadecanoyl chloride |
Specifications
1% max. (as palmitic acid) | |
0.9000g/mL | |
Authentic | |
1.4510 to 1.4530 | |
199.0°C (2.0 mmHg) | |
9.0°C to 14.0°C | |
98% | |
97.5% min. (GC) | |
CH3(CH2)14COCl | |
02, 374 | |
Solubility in water: decomposes. Other solubilities: soluble in ether,decomposes in alcohol | |
CCCCCCCCCCCCCCCC(=O)Cl | |
274.88 | |
274.88 | |
98% |
Palmitoyl chloride | |
160°C | |
Glass bottle | |
0.9 | |
Undesignated | |
500mL | |
112-67-4 | |
C16H31ClO | |
MFCD00000742 | |
palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride | |
ARBOVOVUTSQWSS-UHFFFAOYSA-N | |
hexadecanoyl chloride | |
8206 | |
Liquid |